Learn More
Medchemexpress LLC Methyl 3-hydroxybenzoate | 19438-10-9 | MFCD00002295 | 99.6% | C8H8O3 | 25 G
SDP
Catalog No. 5000358956
Encompass_Preferred
Methyl 3-hydroxybenzoate is a biochemical reagent suitable for life science related research, functioning as a biological material or organic compound. It is a white to off-white solid with a purity of 99.6%.
  • Store powder at -20°C for 3 years
  • Store powder at 4°C for 2 years
  • Store in solvent at -80°C for 2 years
  • Store in solvent at -20°C for 1 year
Catalog No. 50-003-58956 Supplier Medchemexpress LLC Supplier No. 25GHY90004
Methyl 3-hydroxybenzoate is a biochemical reagent suitable for life science related research, functioning as a biological material or organic compound. It is a white to off-white solid with a purity of 99.6%.
  • Store powder at -20°C for 3 years
  • Store powder at 4°C for 2 years
  • Store in solvent at -80°C for 2 years
  • Store in solvent at -20°C for 1 year