Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-hydroxybenzoate, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
19438-10-9
C8H8O3
152.15
MFCD00002295
YKUCHDXIBAQWSF-UHFFFAOYSA-N
methyl m-hydroxybenzoate,3-hydroxybenzoic acid methyl ester,m-carbomethoxyphenol,methyl m-oxybenzoate,m-hydroxybenzoic acid methyl ester,benzoic acid, 3-hydroxy-, methyl ester,3-hydroxy-benzoic acid methyl ester,unii-569m9g3stj,3-methoxycarbonyl phenol,benzoic acid, m-hydroxy-, methyl ester
88068
methyl 3-hydroxybenzoate
COC(=O)C1=CC(=CC=C1)O
19438-10-9
C8H8O3
152.15
MFCD00002295
YKUCHDXIBAQWSF-UHFFFAOYSA-N
methyl m-hydroxybenzoate,3-hydroxybenzoic acid methyl ester,m-carbomethoxyphenol,methyl m-oxybenzoate,m-hydroxybenzoic acid methyl ester,benzoic acid, 3-hydroxy-, methyl ester,3-hydroxy-benzoic acid methyl ester,unii-569m9g3stj,3-methoxycarbonyl phenol,benzoic acid, m-hydroxy-, methyl ester
88068
methyl 3-hydroxybenzoate
COC(=O)C1=CC(=CC=C1)O
Quantity 25g
Color Beige
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 70.0°C to 74.0°C
Boiling Point 280.0°C to 281.0°C (709.0mmHg)
Assay Percent Range 98%
Linear Formula HOC6H4CO2CH3
Formula Weight 152.15
Physical Form Crystals
Percent Purity 98%
Chemical Name or Material Methyl 3-hydroxybenzoate, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 243-071-5