Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-hydroxybenzoate, MP Biomedicals™

Catalog No. MP520249980 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP520249980 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP520249980 Supplier MP Biomedicals, Inc Supplier No. 0520249980

missing translation for 'validMainframeMsg'

Methyl 3-hydroxybenzoate, Off-White Powder

  • Methyl 3-hydroxybenzoate has been used in synthesis of:
  • . 3-hydroxybenzene-1,2-dicarbaldehyde
  • . O-methyl O-[3-methyl-4-(methylthio)phenyl]O-(3-methylcarboxyphenyl) phosphorothioate
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19438-10-9
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00002295
InChI Key YKUCHDXIBAQWSF-UHFFFAOYSA-N
Synonym m-Hydroxybenzoic acid methyl ester, 3-Hydroxy-benzoic Acid Methyl Ester, Methyl 3-hydroxybenzoate
IUPAC Name methyl 3-hydroxybenzoate
SMILES COC(=O)C1=CC=CC(O)=C1

Specifications

Melting Point 70°C to 72°C (literature)
Color Off-white
Boiling Point 280°C to 281°C (709 mmHg) (Literature)
Quantity 100 g
Beilstein 2208129
Solubility Information Soluble in Chloroform, Dimetyl Sulfoxide and Ethanol.
Physical Form Powder
Grade 0.9974
storageNote1 Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.