Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-Hydroxyphenylacetate 98.0+%, TCI America™
SDP

Catalog No. M29251G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29251G 1 g
M29255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M29251G Supplier TCI America Supplier No. M29251G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 42058-59-3
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00017660
InChI Key AMDDOQIUPAINLH-UHFFFAOYSA-N
Synonym 3-Hydroxyphenylacetic Acid Methyl Ester
PubChem CID 39129
IUPAC Name methyl 2-(3-hydroxyphenyl)acetate
SMILES COC(=O)CC1=CC(=CC=C1)O

Specifications

Color Yellow
Boiling Point 146°C
Quantity 1 g
Formula Weight 166.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3-Hydroxyphenylacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.