Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (-)-Methyl ™-3-Hydroxyvalerate

Catalog No. 566551ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-655-1ML 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 56-655-1ML Supplier Sigma Aldrich Supplier No. 566551ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Chiral building block for EPC syntheses; selective anti-alkylation of its dianion; OH protection and reduction of ester group to aldehyde or alcohol functions; stereoselective preparation of a dioxanone with pivalaldehyde

Specifications

CAS 72954-06-4
Melting Point 88°C to 92°C
Molecular Formula C16H13FeP
Refractive Index n20/D 1.427
Linear Formula C5H4PC6H5C5H4Fe
MDL Number MFCD04974069
Quantity 1 g
Synonym 1,1'-(Ferrocenediyl)phenylphosphine
Molecular Weight (g/mol) 292.09 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.