Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl 3-mercaptopropionate

Catalog No. 1089875G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-898-75G 5 g
10-898-7100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-898-75G Supplier Sigma Aldrich Supplier No. 1089875G

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The Progress in Development of Dental Restorative Materials

Specifications

CAS 2935-90-2
Density 1.085 g/mL (at 25°C)
Boiling Point 54°C to 55°C (4 mmHg)
Molecular Formula C4H8O2S
Refractive Index n20/D 1.464 (literature)
Linear Formula HSCH2CH2COOCH3
Quantity 5 g
MDL Number MFCD00004895
Synonym 3-Mercaptopropionic Acid Methyl Ester
Molecular Weight (g/mol) 120.17
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.