Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-mercaptopropionate, 98%

Catalog No. p-165329
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC148870050 5 g Glass bottle
AC148871000 100 g Glass bottle
AC148875000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC148870050 Supplier Thermo Scientific Chemicals Supplier No. 148870050
Only null left
Add to Cart

CAS: 2935-90-2 | C4H8O2S | 120.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2935-90-2
Molecular Formula C4H8O2S
Molecular Weight (g/mol) 120.17
MDL Number MFCD00004895
InChI Key LDTLDBDUBGAEDT-UHFFFAOYSA-N
PubChem CID 18050
IUPAC Name methyl 3-sulfanylpropanoate
SMILES COC(=O)CCS

Specifications

Color Colorless
Density 1.0800g/mL
Boiling Point 166.0°C to 169.0°C
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4620 to 1.4640
Linear Formula HSCH2CH2CO2CH3
Quantity 5 g
Beilstein 03, II, 214
Specific Gravity 1.08
Synonym methyl 3-mercaptopropionate, propanoic acid, 3-mercapto-, methyl ester, 3-mercaptopropionic acid methyl ester, methyl mercaptopropionate, methyl3-mercaptopropionate, methyl 3-mercaptopropanoate, propionic acid, 3-mercapto-, methyl ester, unii-v920f0bcrz, methyl-3-mercaptopropionate, v920f0bcrz
Formula Weight 120.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl 3-mercaptopropionate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Toxic if swallowed.
Harmful in contact with skin.
Fatal if inhaled.
Very
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 220-912-4
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.