Learn More
Medchemexpress LLC Methyl 3-methoxyacrylate | 34846-90-7 | 116.12 | 500 G
SDP
Catalog No. 5000446744
Encompass_Preferred
Methyl 3-methoxyacrylate (3-Methoxyacrylic Acid Methyl Ester) is a Lignans product that can be isolated from the herbs of Lysimachia clethroides. It is for research use only.
  • Can be isolated from the herbs of Lysimachia clethroides
  • For research use only
  • Liquid appearance
  • Colorless to light yellow
  • Store at room temperature for 3 years
  • When in solvent, store at -80°C for 2 years or -20°C for 1 year
Catalog No. 50-004-46744 Supplier Medchemexpress LLC Supplier No. HYW007972500G
Methyl 3-methoxyacrylate (3-Methoxyacrylic Acid Methyl Ester) is a Lignans product that can be isolated from the herbs of Lysimachia clethroides. It is for research use only.
  • Can be isolated from the herbs of Lysimachia clethroides
  • For research use only
  • Liquid appearance
  • Colorless to light yellow
  • Store at room temperature for 3 years
  • When in solvent, store at -80°C for 2 years or -20°C for 1 year