Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-Methoxypropionate 99.0+%, TCI America™
SDP

Catalog No. M0962500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0962500ML 500 mL
M096225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0962500ML Supplier TCI America Supplier No. M0962500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3852-09-3
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.13
MDL Number MFCD00059085
InChI Key BDJSOPWXYLFTNW-UHFFFAOYSA-N
Synonym 3-Methoxypropionic Acid Methyl Ester
PubChem CID 19710
IUPAC Name methyl 3-methoxypropanoate
SMILES COCCC(=O)OC

Specifications

Color Colorless
Boiling Point 144°C
Quantity 500 mL
UN Number 3272
Formula Weight 118.13
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3-Methoxypropionate
EINECSNumber (2)-1350
RTECSNumber UF5274000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.