Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl 3-O-methylgallate | 3934-86-9 | 250 MG
SDP

Catalog No. 5000420669
Encompass_Preferred

Methyl 3-O-methylgallate (M3OMG) is an antioxidant agent that can protect neuronal cells from oxidative damage. It has been shown to mitigate NaF-induced oxidative stress by normalizing TBARS levels, reducing glutathione levels, and restoring diminished antioxidant enzyme activities in rats.

  • Antioxidant agent
  • Protects neuronal cells from oxidative damage
  • Mitigates NaF-induced oxidative stress
  • Molecular weight: 198.17
  • Chemical formula: C9H10O5
  • Appearance: Solid
  • Color: Off-white to yellow
  • Purity: 99.73%
  • Structure classification: Phenols, Polyphenols
  • Initial source: Plants, Rosaceae, Sanguisorba officinalis Linn.

Catalog No. 50-004-20669 Supplier Medchemexpress LLC Supplier No. 250MGHYN6669
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl 3-O-methylgallate (M3OMG) is an antioxidant agent that can protect neuronal cells from oxidative damage. It has been shown to mitigate NaF-induced oxidative stress by normalizing TBARS levels, reducing glutathione levels, and restoring diminished antioxidant enzyme activities in rats.

  • Antioxidant agent
  • Protects neuronal cells from oxidative damage
  • Mitigates NaF-induced oxidative stress
  • Molecular weight: 198.17
  • Chemical formula: C9H10O5
  • Appearance: Solid
  • Color: Off-white to yellow
  • Purity: 99.73%
  • Structure classification: Phenols, Polyphenols
  • Initial source: Plants, Rosaceae, Sanguisorba officinalis Linn.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.