Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl 3-O-methylgallate (M3OMG) | 3934-86-9 | 10 MM 1 ML
SDP

Catalog No. 5000418937
Encompass_Preferred

Methyl 3-O-methylgallate (M3OMG) is an antioxidant agent that can protect neuronal cells from oxidative damage.

  • Antioxidant effect; protects neuronal cells from oxidative damage
  • Mitigates NaF-induced oxidative stress in rat brains by normalizing TBARS levels, reducing glutathione levels, and restoring diminished antioxidant enzyme activities
  • Molecular weight: 198.17
  • Chemical formula: C9H10O5
  • Appearance: solid, off-white to yellow
  • Classified as a phenol and polyphenol
  • Sourced from plants such as Rosaceae and Sanguisorba officinalis Linn.
  • Purity: 99.73%

Catalog No. 50-004-18937 Supplier Medchemexpress LLC Supplier No. HYN666910MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl 3-O-methylgallate (M3OMG) is an antioxidant agent that can protect neuronal cells from oxidative damage.

  • Antioxidant effect; protects neuronal cells from oxidative damage
  • Mitigates NaF-induced oxidative stress in rat brains by normalizing TBARS levels, reducing glutathione levels, and restoring diminished antioxidant enzyme activities
  • Molecular weight: 198.17
  • Chemical formula: C9H10O5
  • Appearance: solid, off-white to yellow
  • Classified as a phenol and polyphenol
  • Sourced from plants such as Rosaceae and Sanguisorba officinalis Linn.
  • Purity: 99.73%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.