Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-oxo-pentanoate, 99+%

Catalog No. AC243165000
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC243165000 500 mL Glass bottle
AC243160250 25 mL Glass bottle
AC243161000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC243165000 Supplier Thermo Scientific Chemicals Supplier No. 243165000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 30414-53-0 | C6H10O3 | 130.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30414-53-0
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD00011705
InChI Key XJMIXEAZMCTAGH-UHFFFAOYSA-N
Synonym methyl 3-oxovalerate, methyl propionylacetate, methyl 3-oxo-pentanoate, pentanoic acid, 3-oxo-, methyl ester, methyl-3-oxo pentanoate, methyl 3-oxo-n-valerate, valeric acid, 3-oxo-, methyl ester, 3-oxovaleric acid methyl ester, 3-oxo-pentanoic acid methyl ester, 3-keto-n-valeric acid methyl ester
PubChem CID 121699
IUPAC Name methyl 3-oxopentanoate
SMILES CCC(=O)CC(=O)OC

Specifications

Melting Point -35°C
Color Colorless to Yellow
Density 1.0300g/mL
Boiling Point 95°C to 97°C (48.0 mmHg)
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Refractive Index 1.4220 to 1.424
Linear Formula CH3CH2COCH2COOCH3
Quantity 500 mL
Fieser 10,415; 01,1022; 04,437; 05,589; 07,324; 11,472; 12,126; 16,303
Specific Gravity 1.03
Solubility Information Solubility in water: slightly soluble. Other solubilities: miscible with most common organic solvents
Formula Weight 130.15
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Methyl 3-oxo-pentanoate, 99+%
Show More Show Less
EINECSNumber 250-184-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.