Learn More
Medchemexpress LLC Methyl 3-oxoheptanoa 10g
Supplier: Medchemexpress LLC HYW09975010G
Methyl 3-oxoheptanoate is the methyl ester of the organic compound 3-oxoheptanoate It has a fruity smell and taste Methyl 3-oxoheptanoate is widely used as a flavoring agent and fragrance in the food and fragrance industry for its pleasant aroma It is also used as an intermediate in the synthesis of other organic compounds such as pharmaceuticals agrochemicals and polymers