Learn More
Medchemexpress LLC Methyl 3-oxohexanoate | 30414-54-1 | 144.17 | 10 G
SDP
Catalog No. 5000445477
Encompass_Preferred
Methyl 3-oxohexanoate (Methyl Butyrylacetate) is an organic compound commonly used in the manufacture of fragrances, drugs, and other chemicals. It functions as a condensation, diazotization, and addition reagent, playing a catalytic role in various organic synthesis reactions and drug synthesis.
  • Used in the food industry for products like juices, beer, and confectionery.
  • Functions as a condensation reagent.
  • Functions as a diazotization reagent.
  • Functions as an addition reagent.
  • Catalytic role in organic synthesis reactions.
  • Useful in drug synthesis.
  • Biochemical reagent for life science-related research.
Catalog No. 50-004-45477 Supplier Medchemexpress LLC Supplier No. HYW00517610G
Methyl 3-oxohexanoate (Methyl Butyrylacetate) is an organic compound commonly used in the manufacture of fragrances, drugs, and other chemicals. It functions as a condensation, diazotization, and addition reagent, playing a catalytic role in various organic synthesis reactions and drug synthesis.
  • Used in the food industry for products like juices, beer, and confectionery.
  • Functions as a condensation reagent.
  • Functions as a diazotization reagent.
  • Functions as an addition reagent.
  • Catalytic role in organic synthesis reactions.
  • Useful in drug synthesis.
  • Biochemical reagent for life science-related research.