Learn More
Medchemexpress LLC Methyl 3-oxohexanoate | 30414-54-1 | 144.17 | 500 G
Supplier: Medchemexpress LLC HYW005176500G
Methyl 3-oxohexanoate (Methyl Butyrylacetate) is an organic compound used in the production of fragrances, drugs, and other chemicals. It functions as a condensation, diazotization, and addition reagent, and acts as a catalyst in certain organic synthesis reactions and drug syntheses. It is also extensively used in the food industry for products such as juices, beer, and confectionery, and serves as a biochemical reagent for life science research.
- Used in the manufacture of fragrances, drugs, and other chemicals
- Functions as a condensation, diazotization, and addition reagent
- Acts as a catalyst in organic synthesis and drug synthesis reactions
- Widely used in the food industry for products like juices, beer, and confectionery
- Biochemical reagent for life science research