Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-phenylpropionate, 98%

Catalog No. AAL0531018
Click to view available options
Quantity:
50 g
250 g

Catalog No. AAL0531018

Supplier: Thermo Scientific Chemicals L0531018

Only null left
Add to Cart

CAS: 103-25-3 | C10H12O2 | 164.204 g/mol

Methyl 3-phenylpropionate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. 3-Phenylpropionic Acid Methyl Ester is an ester derivative of 3-phenylpropionic acid that is sometimes present in fragrance ingredients. It is also used as an application, aromatic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-phenylpropionate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. 3-Phenylpropionic Acid Methyl Ester is an ester derivative of 3-phenylpropionic acid that is sometimes present in fragrance ingredients. It is also used as an application, aromatic intermediate.

Solubility
Soluble in chloroform. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-25-3
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00017209
InChI Key RPUSRLKKXPQSGP-UHFFFAOYSA-N
Synonym methyl 3-phenylpropionate, methyl hydrocinnamate, benzenepropanoic acid, methyl ester, 3-phenylpropionic acid methyl ester, methyl benzenepropanoate, hydrocinnamic acid, methyl ester, methyl dihydrocinnamate, methyl beta-phenylpropionate, methyl b-phenylpropionate, unii-111lc1gi10
PubChem CID 7643
IUPAC Name methyl 3-phenylpropanoate
SMILES COC(=O)CCC1=CC=CC=C1

Specifications

Density 1.045
Boiling Point 236°C to 239°C
Flash Point 100°C (212°F)
Refractive Index 1.502
Quantity 50 g
Beilstein 1909591
Solubility Information Soluble in chloroform. Insoluble in water.
Formula Weight 164.2
Percent Purity 98%
Chemical Name or Material Methyl 3-phenylpropionate
EINECSNumber 203-092-2
TSCA Yes

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.