Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-(trimethylsilyloxy)crotonate, 90%, mixture of cis and trans, Thermo Scientific™

Catalog No. AC336340100
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC336340100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC336340100 Supplier Thermo Scientific Chemicals Supplier No. 336340100

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26767-00-0
Molecular Formula C8H16O3Si
Molecular Weight (g/mol) 188.298
InChI Key OQNKCUVOGBTGDJ-VOTSOKGWSA-N
Synonym methyl 3-trimethylsiloxy crotonate, methyl 2e-3-trimethylsilyl oxy but-2-enoate, methyl-3-trimethylsiloxy crotonate, methyl 3-trimethylsilyl oxy but-2-enoate, methyl3-trimethylsilyloxy crotonate, methyl 3-trimethylsiloxy-2-butenoate, methyl e-3-trimethylsilyloxybut-2-enoate, methyl acetoacetate enol trimethylsilylether, 3-trimethylsiloxy-2-butenoic acid methyl ester, e-methyl 3-trimethylsilyl oxy but-2-enoate
PubChem CID 6115690
IUPAC Name methyl (E)-3-trimethylsilyloxybut-2-enoate
SMILES CC(=CC(=O)OC)O[Si](C)(C)C

Specifications

Color Colorless to Yellow
Density 0.94g/mL
Infrared Spectrum Authentic
Assay Percent Range 90%
Linear Formula (CH3)3SiOC(CH3)CHCOOCH3
Quantity 10 g
Formula Weight 188.3
Percent Purity ≥88%
Grade Technical
Physical Form Liquid
Chemical Name or Material Methyl 3-(trimethylsilyloxy)crotonate, techn., 90%
Show More Show Less
Hazard Category MOISTURE SENSITIVE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.