Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-(1H-pyrazol-1-ylmethyl)benzoate, 97%, Thermo Scientific™

Catalog No. cc56022da
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC56022DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC56022DA Supplier Thermo Scientific Chemicals Supplier No. CC56022DA
Only null left
Add to Cart

Chemical Identifiers

CAS 397328-86-8
Molecular Formula C12H12N2O2
Molecular Weight (g/mol) 216.24
MDL Number MFCD08435918
InChI Key VMCQOZFWPDOGLG-UHFFFAOYSA-N
Synonym methyl 4-1h-pyrazol-1-ylmethyl benzoate, methyl 4-pyrazol-1-ylmethyl benzoate, methyl 4-1h-pyrazol-1-yl methyl benzoate, methyl 4-pyrazolylmethyl benzoate, benzoic acid,4-1h-pyrazol-1-ylmethyl-, methyl ester
PubChem CID 23510322
IUPAC Name methyl 4-(pyrazol-1-ylmethyl)benzoate
SMILES COC(=O)C1=CC=C(C=C1)CN2C=CC=N2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 216.24
Percent Purity 97%
Chemical Name or Material Methyl 4-(1H-pyrazol-1-ylmethyl)benzoate
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.