Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-[2-(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoate 98.0+%, TCI America™
SDP

Catalog No. M24261G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24261G 1 g
M24265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24261G Supplier TCI America Supplier No. M24261G

Chemical Identifiers

CAS 155405-80-4
Molecular Formula C16H16N4O3
Molecular Weight (g/mol) 312.329
MDL Number MFCD17976681
InChI Key GDEKSBCRJKAARB-UHFFFAOYSA-N
Synonym 4-[2-(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoic Acid Methyl Ester, 2-Amino-4,7-dihydro-5-[2-[4-(methoxycarbonyl)phenyl]ethyl]-4-oxo-3H-pyrrolo[2,3-d]pyrimidine
PubChem CID 11023369
IUPAC Name methyl 4-[2-(2-amino-4-oxo-1,7-dihydropyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoate
SMILES COC(=O)C1=CC=C(C=C1)CCC2=CNC3=C2C(=O)N=C(N3)N

Specifications

Quantity 1 g
Formula Weight 312.33
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-[2-(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.