Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl 4-[4-[3-(4-methylphenyl)sulfonyloxypropyl]triazol-1-yl]benzoate | 2171519-89-2 | >98.0% | C20H21N3O5S | 10MG
SDP

Catalog No. 5000259780
Encompass_Preferred

Cjoc42 is a small-molecule research compound that binds gankyrin and modulates the MDM2/p53 pathway, restoring p53-dependent transcription and sensitizing cells to DNA damage. It is provided as a high-purity, DMSO-soluble reagent intended for biochemical and cell-based oncology studies.

  • Binds gankyrin and inhibits its activity in a dose-dependent manner.
  • Restores p53-dependent transcription and increases sensitivity to DNA damage.
  • Suitable for biochemical and cell-based assays.
  • High purity (>98.0%) for reliable experimental results.
  • Soluble in DMSO for easy stock solution preparation.
  • Recommended storage at 4 °C to maintain stability.

Catalog No. 50-002-59780 Supplier Medchemexpress LLC Supplier No. HY13805410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cjoc42 is a small-molecule research compound that binds gankyrin and modulates the MDM2/p53 pathway, restoring p53-dependent transcription and sensitizing cells to DNA damage. It is provided as a high-purity, DMSO-soluble reagent intended for biochemical and cell-based oncology studies.

  • Binds gankyrin and inhibits its activity in a dose-dependent manner.
  • Restores p53-dependent transcription and increases sensitivity to DNA damage.
  • Suitable for biochemical and cell-based assays.
  • High purity (>98.0%) for reliable experimental results.
  • Soluble in DMSO for easy stock solution preparation.
  • Recommended storage at 4 °C to maintain stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.