Learn More
Medchemexpress LLC Methyl (4-((4-fluorobenzyl)(prop-2-yn-1-yl)amino)-2,6-dimethylphenyl)carbamate | 1821222-10-9 | MFCD34676457 | 99.5% | C20H21FN2O2 | 10MG
SDP
Catalog No. 5000260913
Encompass_Preferred
HN37 is a small-molecule KCNQ2 (Kv7) activator intended for laboratory research use. It exhibits potent neuronal Kv7 activation with reported antiepileptic activity (EC50 ≈ 37 nM) and contains an alkyne functional group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) for bioconjugation and probe synthesis. The compound is provided as a high-purity solid or DMSO solution for experimental use.
  • Potent KCNQ2 (Kv7) activator with EC50 ≈ 37 nM.
  • Contains an alkyne group for CuAAC click chemistry.
  • High purity (99.53%) suitable for research applications.
  • Supplied in solid and DMSO solution formats.
  • Intended for laboratory research only; not for human use.
Catalog No. 50-002-60913 Supplier Medchemexpress LLC Supplier No. HY14501610MG
HN37 is a small-molecule KCNQ2 (Kv7) activator intended for laboratory research use. It exhibits potent neuronal Kv7 activation with reported antiepileptic activity (EC50 ≈ 37 nM) and contains an alkyne functional group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) for bioconjugation and probe synthesis. The compound is provided as a high-purity solid or DMSO solution for experimental use.
  • Potent KCNQ2 (Kv7) activator with EC50 ≈ 37 nM.
  • Contains an alkyne group for CuAAC click chemistry.
  • High purity (99.53%) suitable for research applications.
  • Supplied in solid and DMSO solution formats.
  • Intended for laboratory research only; not for human use.