Learn More
Medchemexpress LLC Methyl 4-[6-[4-(methoxycarbonylamino)phenyl]-4-morpholin-4-ylpyrazolo[3,4-d]pyrimidin-1-yl] | 1062169-56-5 | 98.0% | 25 MG
SDP
Catalog No. 5000234376
Encompass_Preferred
WYE-354 is a cell-permeable, ATP-competitive inhibitor of mechanistic target of rapamycin (mTOR) used as a research tool to block signaling through mTORC1 and mTORC2 in cell signaling and oncology studies.
  • Potent mTOR inhibition with IC50 ≈ 4-5 nM.
  • Cell-permeable small molecule suitable for cellular assays.
  • High purity (98.0%) for reproducible results.
  • Molecular weight 495.53 g/mol; formula C24H29N7O5.
  • White to off-white solid; soluble in DMSO; store powder at -20°C.
Catalog No. 50-002-34376 Supplier Medchemexpress LLC Supplier No. HY1203425MG
WYE-354 is a cell-permeable, ATP-competitive inhibitor of mechanistic target of rapamycin (mTOR) used as a research tool to block signaling through mTORC1 and mTORC2 in cell signaling and oncology studies.
  • Potent mTOR inhibition with IC50 ≈ 4-5 nM.
  • Cell-permeable small molecule suitable for cellular assays.
  • High purity (98.0%) for reproducible results.
  • Molecular weight 495.53 g/mol; formula C24H29N7O5.
  • White to off-white solid; soluble in DMSO; store powder at -20°C.