Learn More
Medchemexpress LLC Methyl 4-[8-(hydroxyamino)-8-oxooctanoyl]oxybenzoate | 946150-57-8 | MFCD28986227 | 98.0% | 323.34 | C16H21NO6 | 25 MG
SDP
Catalog No. 5000213127
Encompass_Preferred
Remetinostat is a hydroxamic acid-based histone deacetylase (HDAC) inhibitor supplied as a white to off-white solid for laboratory research. It has demonstrated efficacy in imiquimod-induced models of psoriatic dermatitis and is used in preclinical studies of cutaneous T-cell lymphoma. The compound is supplied with defined purity and storage recommendations to support reproducible experiments.
  • Hydroxamic acid-based histone deacetylase (HDAC) inhibitor.
  • Validated activity in psoriatic dermatitis animal models.
  • Applicable to studies of cutaneous T-cell lymphoma mechanisms.
  • High purity suitable for research (98.0%).
  • Stable solid form with recommended cold storage for prolonged shelf life.
Catalog No. 50-002-13127 Supplier Medchemexpress LLC Supplier No. HY10036525MG
Remetinostat is a hydroxamic acid-based histone deacetylase (HDAC) inhibitor supplied as a white to off-white solid for laboratory research. It has demonstrated efficacy in imiquimod-induced models of psoriatic dermatitis and is used in preclinical studies of cutaneous T-cell lymphoma. The compound is supplied with defined purity and storage recommendations to support reproducible experiments.
  • Hydroxamic acid-based histone deacetylase (HDAC) inhibitor.
  • Validated activity in psoriatic dermatitis animal models.
  • Applicable to studies of cutaneous T-cell lymphoma mechanisms.
  • High purity suitable for research (98.0%).
  • Stable solid form with recommended cold storage for prolonged shelf life.