Learn More
Sigma Aldrich methyl 4-amino-3-hydroxybenzoate
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Benzaldehyde dimethyl acetal is an organic building block
- Pyridinium tosylate-catalyzed acetal exchange reaction between benzaldehyde dimethyl acetal and 6-o-(tert-butyldiphenylsilyl)-1,2-o-isopropylidene-α-d-glucofuranose is reported to afford 3,5-o-benzylidene-1,2-o-isopropylidene-α-d-glucofuranose
- The kinetics of the hydrolysis of benzaldehyde dimethyl acetal over amberlite IR-120 has been studied using a circulated batch reactor in dioxane
- One-pot tandem conversion of benzaldehydedimethylacetal to trans-1-nitro-2-phenylethylene has been reported.benzaldehyde dimethyl acetal is suitable for use in the synthesis of 4,6-dihydroxy sugar, required for the total synthesis of porphyromonas gingivalis 381 derived lipid a
- It may be used in the preparation of 1-o-methyl-2,3-di-o-galloyl-α-d-glucose
Specifications
Specifications
| Molecular Formula | C8H9NO3 |
| Linear Formula | C8H9NO3 |
| MDL Number | MFCD00017093 |
| Quantity | 100 mL |
| Molecular Weight (g/mol) | 167.16 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.