Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-aminonicotinate, 98%, Thermo Scientific Chemicals

Catalog No. p-7028168
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2532406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2532406 Supplier Thermo Scientific Chemicals Supplier No. B2532406
Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16135-36-7
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00955996
InChI Key OCARFFAPQGYGBP-UHFFFAOYSA-N
Synonym methyl 4-aminonicotinate, 4-aminonicotinic acid methyl ester, 4-amino-nicotinic acid methyl ester, 4-aminopyridine-3-carboxylic acid methyl ester, 3-pyridinecarboxylic acid, 4-amino-, methyl ester, nicotinic acid, 4-amino-, methyl ester, 4-aminonicotinicacidmethylester, methyl-4-aminonicotinate, ksc493a3l, tpc-py097
PubChem CID 592353
IUPAC Name methyl 4-aminopyridine-3-carboxylate
SMILES COC(=O)C1=C(N)C=CN=C1

Specifications

Melting Point 172°C to 174°C
Quantity 5 g
Formula Weight 152.15
Percent Purity 98%
Chemical Name or Material Methyl 4-aminonicotinate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.