Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl (+)-4-Boc-piperazine-2-carboxylate

Catalog No. AAH2788603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2788603 1 g
AAH2788606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2788603 Supplier Thermo Scientific Chemicals Supplier No. H2788603
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 129799-08-2 | C11H21N2O4 | 245.30 g/mol

It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129799-08-2
Molecular Formula C11H21N2O4
Molecular Weight (g/mol) 245.30
MDL Number MFCD01632465,MFCD04115325,MFCD04115326
InChI Key QUKAHFCVKNRRBU-QMMMGPOBSA-O
Synonym methyl 4-boc-piperazine-2-carboxylate, 1-tert-butyl 3-methyl piperazine-1,3-dicarboxylate, n-4-boc-2-piperazinecarboxylic acid methyl ester, 1-n-boc-piperazine-3-carboxylic acid methyl ester, 4n-boc-piperazine-2-carboxylic acid methyl ester, piperazine-1,3-dicarboxylic acid 1-tert-butyl ester 3-methyl ester, 1,3-piperazinedicarboxylic acid, 1-1,1-dimethylethyl 3-methyl ester, 1-n-boc-piperazine-3-carboxylic acid methyl ester; piperazine-1,3-dicarboxylic acid 1-tert-butyl ester 3-methyl ester, tert-butyl 3-methoxycarbonyl piperazinecarboxylate, pubchem18338
PubChem CID 2756819
IUPAC Name 1-O-tert-butyl 3-O-methyl piperazine-1,3-dicarboxylate
SMILES COC(=O)[C@@H]1CN(CC[NH2+]1)C(=O)OC(C)(C)C

Specifications

Quantity 1 g
Solubility Information Soluble in dimethyl sulfoxide.
Formula Weight 244.29
Chemical Name or Material Methyl (+/-)-4-Boc-piperazine-2-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.