Learn More
Medchemexpress LLC Methyl 4-boronobenzoate | 99768-12-4 | 179.97 | 25 G
SDP
Catalog No. 5000448124
Encompass_Preferred
Methyl 4-boronobenzoate, also known as 4-Methoxycarbonylbenzeneboronic acid, is a biochemical reagent used as a biological material or organic compound in life science research. It should be stored as powder at -20°C for 3 years or 4°C for 2 years. In solvent, store at -80°C for 6 months or -20°C for 1 month.
  • Used in tandem-type Pd(II)-catalyzed oxidative Heck reactions.
  • A reagent for intramolecular CH amidation sequences.
  • Employed in copper-mediated ligandless aerobic fluoroalkylation of arylboronic acids.
  • Applicable in one-pot ipso-nitration of arylboronic acids.
  • Used in palladium-phosphine-catalyzed Suzuki-Miyaura coupling.
  • Employed in the preparation of biaryls via nickel-catalyzed Suzuki-Miyaura cross-coupling.
Catalog No. 50-004-48124 Supplier Medchemexpress LLC Supplier No. 25GHYW003977
Methyl 4-boronobenzoate, also known as 4-Methoxycarbonylbenzeneboronic acid, is a biochemical reagent used as a biological material or organic compound in life science research. It should be stored as powder at -20°C for 3 years or 4°C for 2 years. In solvent, store at -80°C for 6 months or -20°C for 1 month.
  • Used in tandem-type Pd(II)-catalyzed oxidative Heck reactions.
  • A reagent for intramolecular CH amidation sequences.
  • Employed in copper-mediated ligandless aerobic fluoroalkylation of arylboronic acids.
  • Applicable in one-pot ipso-nitration of arylboronic acids.
  • Used in palladium-phosphine-catalyzed Suzuki-Miyaura coupling.
  • Employed in the preparation of biaryls via nickel-catalyzed Suzuki-Miyaura cross-coupling.