Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-Bromo-2-(bromomethyl)benzoate 97.0+%, TCI America™
SDP

Catalog No. M2941200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2941200MG 200 mg
M29411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2941200MG Supplier TCI America Supplier No. M2941200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 78471-43-9
Molecular Formula C9H8Br2O2
Molecular Weight (g/mol) 307.969
MDL Number MFCD04114319
InChI Key SGFACFBLUAWICV-UHFFFAOYSA-N
Synonym methyl 4-bromo-2-bromomethyl benzoate, methyl 4-bromo-2-bromomethyl-benzoate, 4-bromo-2-bromomethylbenzoic acid methyl ester, 4-bromo-2-bromomethyl-benzoic acid methyl ester, benzoic acid, 4-bromo-2-bromomethyl-, methyl ester, 4-bromo-2-bromomethyl benzoic acid methyl ester, acmc-209pec, methyl alpha,5-dibromo-o-toluate, methyl 4bromo-2-bromomethylbenzoate, methyl4-bromo-2-bromomethylbenzoate
PubChem CID 22031079
IUPAC Name methyl 4-bromo-2-(bromomethyl)benzoate
SMILES COC(=O)C1=C(C=C(C=C1)Br)CBr

Specifications

Melting Point 81°C
Quantity 200 mg
UN Number 3261
Formula Weight 307.97
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Bromo-2-(bromomethyl)benzoate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.