Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4'-bromobiphenyl-3-carboxylate, 95%, Thermo Scientific™

Catalog No. AAH5190506
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5190506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5190506 Supplier Thermo Scientific Chemicals Supplier No. H5190506

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149506-25-2
Molecular Formula C14H11BrO2
Molecular Weight (g/mol) 291.144
MDL Number MFCD05980426
InChI Key UTOKFKAMZICCBU-UHFFFAOYSA-N
Synonym 4'-bromo-biphenyl-3-carboxylic acid methyl ester, 4'-bromobiphenyl-3-carboxylic acid methyl ester, methyl 3-4-bromophenyl benzoate, methyl 4'-bromobiphenyl-3-carboxylate, methyl 4'-bromo-1,1'-biphenyl-3-carboxylate, amtda029, 4-bromo-3'-methoxycarbonyl biphenyl, methyl 4'-bromo 1,1'-biphenyl-3-carboxylate, 4\'-bromo-biphenyl-3-carboxylic acid methyl ester
PubChem CID 23005260
IUPAC Name methyl 3-(4-bromophenyl)benzoate
SMILES COC(=O)C1=CC=CC(=C1)C2=CC=C(C=C2)Br

Specifications

Quantity 5 g
Formula Weight 291.14
Percent Purity 95%
Chemical Name or Material Methyl 4'-bromobiphenyl-3-carboxylate
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.