Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific Methyl 4-(bromomethyl)-3-methoxybenzoate, 70264-94-7, MFCD00270115, 5g

Catalog No. 501472548
Encompass_Preferred

Molecular Formula C10H11BrO3, Molecular Weight 259.11, Melting Point 87-89°

Catalog No. 50-147-2548 Supplier Matrix Scientific Supplier No. 0010885G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Molecular Formula C10H11BrO3, Molecular Weight 259.11, Melting Point 87-89°

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.