Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-Chloro-2-methylphenoxyacetate 94.0+%, TCI America™
SDP

Catalog No. C059825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C059825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C059825G Supplier TCI America Supplier No. C059825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2436-73-9
Molecular Formula C10H11ClO3
Molecular Weight (g/mol) 214.645
MDL Number MFCD00055259
InChI Key VWERIRLJUWTNDA-UHFFFAOYSA-N
Synonym mcpa methyl ester, mcpa-methyl, methyl 4-chloro-2-methylphenoxyacetate, methyl 2-4-chloro-2-methylphenoxy acetate, mcpa-methyl iso, acetic acid, 4-chloro-2-methylphenoxy-, methyl ester, unii-cm4042hz0o, methyl 4-chloro-2-methylphenoxy acetate, 2-methyl-4-chlorophenoxyacetic acid methyl ester, 4-chloro-2-methylphenoxy acetic acid methyl ester
PubChem CID 17089
IUPAC Name methyl 2-(4-chloro-2-methylphenoxy)acetate
SMILES CC1=C(C=CC(=C1)Cl)OCC(=O)OC

Specifications

Color Yellow
Boiling Point 127°C
Quantity 25 g
Formula Weight 214.65
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 4-Chloro-2-methylphenoxyacetate
RTECSNumber AG2290000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.