Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl 4-chloro-4-oxobutyrate

Catalog No. C11049100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C11049100G 100 g
C1104925G 25 g
C110495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C11049100G Supplier Sigma Aldrich Supplier No. C11049100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methyl 4-chloro-4-oxobutyrate

Specifications

CAS 1490-25-1
Density 1.223 g/mL (at 25°C (literature))
Boiling Point 58°C to 65°C (3 mmHg)
Molecular Formula C5H7ClO3
Refractive Index n20/D 1.44 (literature)
Linear Formula CH3OCOCH2CH2COCl
MDL Number MFCD00000750
Quantity 100 g
Synonym Methyl succinyl chloride; 3-(Carbomethoxy)propionyl chloride; Methyl 3-(chloroformyl)propionate; Succinic acid monomethylester chloride
Molecular Weight (g/mol) 150.56
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.