Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-Fluorobenzoate 97.0+%, TCI America™
SDP

Catalog No. F0403500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0403500G 500 g
F040325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0403500G Supplier TCI America Supplier No. F0403500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 403-33-8
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00017959
InChI Key MSEBQGULDWDIRW-UHFFFAOYSA-N
Synonym methyl p-fluorobenzoate, 4-fluorobenzoic acid methyl ester, methyl 4-fluorobenzoic acid, benzoic acid, 4-fluoro-, methyl ester, methyl4-fluorobenzoate, methyl-4-fluorobenzoate, 4-fluoro-benzoic acid methyl ester, pubchem3566, acmc-1aef0, asischem p32019
PubChem CID 67878
IUPAC Name methyl 4-fluorobenzoate
SMILES COC(=O)C1=CC=C(C=C1)F

Specifications

Color Yellow
Boiling Point 197°C
Quantity 500 g
Formula Weight 154.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 4-Fluorobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.