Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-fluorophenylacetate, 99%, Thermo Scientific Chemicals

Catalog No. AC304671000
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC304671000 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC304671000 Supplier Thermo Scientific Chemicals Supplier No. 304671000

missing translation for 'validMainframeMsg'

CAS: 34837-84-8 | Molecular Formula: C9H9FO2 | Molecular Weight: 168.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34837-84-8
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.06
InChI Key AJPPKGMEHMXPMC-UHFFFAOYSA-N
Synonym methyl 2-4-fluorophenyl acetate, methyl 4-fluorophenylacetate, 4-fluorophenylacetic acid methyl ester, pubchem24389, acmc-209iaq, methyl-4-fluorphenylacetate, methyl-4-fluorophenylacetate, methyl 4-fluorophenyl-acetate, methyl 4-fluorophenyl acetate
PubChem CID 2733233
IUPAC Name methyl 2-(4-fluorophenyl)acetate
SMILES COC(=O)CC1=CC=C(C=C1)F

Specifications

Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4860 to 1.4880
Linear Formula FC6H4CH2CO2CH3
Quantity 100 g
Formula Weight 168.06
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl 4-fluorophenylacetate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.