Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-formyl-1-methyl-1H-pyrrole-2-carboxylate, 95%, Thermo Scientific™

Catalog No. SEW06271DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SEW06271DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SEW06271DA Supplier Thermo Scientific Chemicals Supplier No. SEW06271DA
Add to Cart

Chemical Identifiers

CAS 67858-47-3
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD01571196
InChI Key GXSZDESAIZEFGZ-UHFFFAOYSA-N
Synonym methyl 4-formyl-1-methyl-1h-pyrrole-2-carboxylate, pubchem12439, methylformylmethylpyrrolecarboxylate, 1h-pyrrole-2-carboxylicacid, 4-formyl-1-methyl-, methyl ester, 4-formyl-1-methyl-1h-pyrrole-2-carboxylic acid methyl ester
PubChem CID 2798349
IUPAC Name methyl 4-formyl-1-methylpyrrole-2-carboxylate
SMILES CN1C=C(C=C1C(=O)OC)C=O

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 167.16
Percent Purity 95%
Chemical Name or Material Methyl 4-formyl-1-methyl-1H-pyrrole-2-carboxylate
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.