Learn More
Medchemexpress LLC Methyl 4-hydroxy-3-methoxycinnamate | 2309-07-1 | MFCD00017208 | 99.9% | 208.21 g·mol⁻¹ | C11H12O4 | 500 MG
SDP
Catalog No. 5000301024
Encompass_Preferred
Ferulic acid methyl ester (methyl ferulate) is the methyl ester derivative of ferulic acid (CAS 2309-07-1). It is used as a research reagent with reported antioxidant and anti-inflammatory activity and has been reported to affect p38 MAPK; supplied for biochemical and cellular studies at high purity.
  • Methyl ester derivative of ferulic acid.
  • Reported antioxidant and anti-inflammatory activity.
  • Reported activity in p38 MAPK assays.
  • High purity suitable for biochemical and cellular research.
  • Soluble in common organic solvents such as DMSO.
  • Available in small research pack sizes (for example, 500 mg).
Catalog No. 50-003-01024 Supplier Medchemexpress LLC Supplier No. HYW018643500MG
Ferulic acid methyl ester (methyl ferulate) is the methyl ester derivative of ferulic acid (CAS 2309-07-1). It is used as a research reagent with reported antioxidant and anti-inflammatory activity and has been reported to affect p38 MAPK; supplied for biochemical and cellular studies at high purity.
  • Methyl ester derivative of ferulic acid.
  • Reported antioxidant and anti-inflammatory activity.
  • Reported activity in p38 MAPK assays.
  • High purity suitable for biochemical and cellular research.
  • Soluble in common organic solvents such as DMSO.
  • Available in small research pack sizes (for example, 500 mg).