Learn More
Selleck Chemical LLC Methyl 4-hydroxybenzoate S3985-25mg
SDP
Catalog No. 503108398
Encompass_Preferred
Methyl 4-hydroxybenzoate (Methylparaben Methyl p-hydroxybenzoate Nipagin) is an anti-fungal agent often used in a variety of cosmetics personal-care products and food preservatives
Catalog No. 50-310-8398 Supplier Selleck Chemical LLC Supplier No. S398525MG
Methyl 4-hydroxybenzoate (Methylparaben Methyl p-hydroxybenzoate Nipagin) is an anti-fungal agent often used in a variety of cosmetics personal-care products and food preservatives