Learn More
Medchemexpress LLC Methyl 4-hydroxybenzoate-d4 | 362049-51-2 | 156.17 | 5 MG
SDP
Catalog No. 5000390064
Encompass_Preferred
Methyl paraben-d4 is the deuterium-labeled form of methyl paraben, which is the methyl ester of p-hydroxybenzoic acid. Isolated from the barks of Tsuga dumosa, methyl paraben is a standardized chemical allergen. It serves as a stable, non-volatile antimicrobial preservative in various products. Its physiological effect involves increased histamine release and cell-mediated immunity.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes incorporated into drug molecules largely for quantitation during drug development.
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-90064 Supplier Medchemexpress LLC Supplier No. HYN0349S50MG
Methyl paraben-d4 is the deuterium-labeled form of methyl paraben, which is the methyl ester of p-hydroxybenzoic acid. Isolated from the barks of Tsuga dumosa, methyl paraben is a standardized chemical allergen. It serves as a stable, non-volatile antimicrobial preservative in various products. Its physiological effect involves increased histamine release and cell-mediated immunity.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes incorporated into drug molecules largely for quantitation during drug development.
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs.