Learn More
Medchemexpress LLC Methyl 4-hydroxyphenylacetate | 14199-15-6 | 166.18 | 100 G
SDP
Catalog No. 5000449020
Encompass_Preferred
Methyl 4-hydroxyphenylacetate is a natural compound that functions as a methyl ester, formed through the formal condensation of the carboxy group of 4-Hydroxyphenylacetic acid with methanol. It is isolated from microorganisms, specifically the mangrove-derived fungus Eupenicillium sp. HJ002.
  • Natural compound
  • Colorless to light yellow liquid
  • Density: 1.1708 g/cm³
  • Derived from microorganisms
Catalog No. 50-004-49020 Supplier Medchemexpress LLC Supplier No. HYW001084100G
Methyl 4-hydroxyphenylacetate is a natural compound that functions as a methyl ester, formed through the formal condensation of the carboxy group of 4-Hydroxyphenylacetic acid with methanol. It is isolated from microorganisms, specifically the mangrove-derived fungus Eupenicillium sp. HJ002.
  • Natural compound
  • Colorless to light yellow liquid
  • Density: 1.1708 g/cm³
  • Derived from microorganisms