Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl 4-hydroxyphenylacetate | 14199-15-6 | 166.18 | 50 G
SDP

Catalog No. 5000449557
Encompass_Preferred

Methyl 4-hydroxyphenylacetate is a natural compound, a methyl ester formed by the condensation of 4-Hydroxyphenylacetic acid with methanol. It presents as a colorless to light yellow liquid with a density of 1.1708 g/cm³. This compound is classified as a phenol and monophenol, originally isolated from the mangrove-derived fungus Eupenicillium sp. HJ002.

  • Natural compound and methyl ester
  • Colorless to light yellow liquid appearance
  • Density of 1.1708 g/cm³
  • Classified as a phenol and monophenol
  • Isolated from specific mangrove-derived fungus
  • Intended for research use only
  • Storage at 4°C under nitrogen for optimal stability
  • Recommended shipping at room temperature (within continental US)
  • Stock solutions stable at -80°C for up to 6 months or -20°C for 1 month under nitrogen

Catalog No. 50-004-49557 Supplier Medchemexpress LLC Supplier No. HYW00108450G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Methyl 4-hydroxyphenylacetate is a natural compound, a methyl ester formed by the condensation of 4-Hydroxyphenylacetic acid with methanol. It presents as a colorless to light yellow liquid with a density of 1.1708 g/cm³. This compound is classified as a phenol and monophenol, originally isolated from the mangrove-derived fungus Eupenicillium sp. HJ002.

  • Natural compound and methyl ester
  • Colorless to light yellow liquid appearance
  • Density of 1.1708 g/cm³
  • Classified as a phenol and monophenol
  • Isolated from specific mangrove-derived fungus
  • Intended for research use only
  • Storage at 4°C under nitrogen for optimal stability
  • Recommended shipping at room temperature (within continental US)
  • Stock solutions stable at -80°C for up to 6 months or -20°C for 1 month under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.