Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-iodopyrrole-2-carboxylate, 97%, Thermo Scientific™

Catalog No. AAH33193MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH33193MD 250 mg
AAH3319303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH33193MD Supplier Thermo Scientific Chemicals Supplier No. H33193MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40740-41-8
Molecular Formula C6H6INO2
Molecular Weight (g/mol) 251.023
MDL Number MFCD07368824
InChI Key KRNGDJYQOKCQCD-UHFFFAOYSA-N
PubChem CID 11673249
IUPAC Name methyl 4-iodo-1H-pyrrole-2-carboxylate
SMILES COC(=O)C1=CC(=CN1)I

Specifications

Melting Point 76°C to 78°C
Color White to Brown
Quantity 250 mg
Sensitivity Light sensitive
Synonym methyl 4-iodopyrrole-2-carboxylate, 4-iodopyrrole-2-carboxylic acid methyl ester, 1h-pyrrole-2-carboxylic acid, 4-iodo-, methyl ester, 4-iodo-1h-pyrrole-2-carboxylic acid methyl ester, methyliodopyrrolecarboxylate, 4-iodopyrrole-2-carboxylicacidmethylester, 4-iodo-2-methoxycarbonylpyrrole, methyl4-iodopyrrole-2-carboxylate, methyl 4-iodo-2-pyrrolecarboxylate, methyl 4-iodo-1h-pyrrole-2-carboxylate
Formula Weight 251
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 4-iodopyrrole-2-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.