Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl 4-oxobutanoate

Catalog No. 5890981G
Encompass
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
58-909-81G 1 g
50-290-8034 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 58-909-81G Supplier Sigma Aldrich Supplier No. 5890981G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methyl 4-oxobutanoate may be used as a starting material to synthesize (-)-deoxoprosophylline, a piperidine alkaloid with therapeutic potential.

Specifications

CAS 13865-19-5
Density 1.109 g/mL (at 25°C (literature))
Boiling Point 187°C to 188°C (lit.)
Molecular Formula C5H8O3
Refractive Index n20/D 1.424 (literature)
Linear Formula H3COCOCH2CH2CHO
Quantity 1 g
MDL Number MFCD00082185
Molecular Weight (g/mol) 116.12
Percent Purity 90%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.