Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl 4-pentenoate

Catalog No. 496611GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-661-1GF 1 g
49-661-5GF 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 49-661-1GF Supplier Sigma Aldrich Supplier No. 496611GF
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Methyl 4-pentenoate is an unsaturated methyl ester that is prepared by the esterification of 1-pentenoic acid with methanol
  • It undergoes amidation with formamide via acetone-initiated photochemical reaction to form a 1:1 adduct
  • The metathesis of methyl 4-pentenoate with different mo(vi)alkylidene complexes have been reported
  • It acts as a chain transfer agent during the polymerization of exo,exo-5,6-bis(methoxymethyl)-7-oxabicyclo[2.2.1]hept-2-ene using ruii(h20)6(tos)2 (tos =p-toluenesulfonate) as a catalyst

Specifications

CAS 818-57-5
Molecular Formula C6H10O2
Refractive Index n20/D 1.415; n20/D 1.415
Linear Formula C6H10O2
MDL Number MFCD03990590
Quantity 1 g
Synonym 4-Pentenoic acid methyl ester; Allylacetic acid methyl ester; Methyl allylacetate
Molecular Weight (g/mol) 114.14
Percent Purity ≥95.0% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.