Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate, 95%, Thermo Scientific™

Catalog No. SEW02756DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SEW02756DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SEW02756DA Supplier Thermo Scientific Chemicals Supplier No. SEW02756DA
Add to Cart

Chemical Identifiers

CAS 237385-98-7
Molecular Formula C13H9F3O2S
Molecular Weight (g/mol) 286.268
MDL Number MFCD00110251
InChI Key RDMSDMSSVFMCKQ-UHFFFAOYSA-N
Synonym methyl 4-phenyl-5-trifluoromethyl thiophene-2-carboxylate, methyl 4-phenyl-5-trifluoromethyl-thiophene-2-carboxylate, 2-thiophenecarboxylicacid, 4-phenyl-5-trifluoromethyl-, methyl ester, 2-methoxycarbonyl-4-phenyl-5-trifluoromethyl thiophene, methyl 4-phenyl-5-trifluoromethyl-2-thiophenecarboxylate, methyl 4-phenyl-5-trifluoromethyl thiophene-2carboxylate, 4-phenyl-5-trifluoromethyl-2-thiophenecarboxylic acid methyl ester, 4-phenyl-5-trifluoromethyl-thiophene-2-carboxylic acid methyl ester
PubChem CID 2781442
IUPAC Name methyl 4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate
SMILES COC(=O)C1=CC(=C(S1)C(F)(F)F)C2=CC=CC=C2

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 286.27
Percent Purity 95%
Chemical Name or Material Methyl 4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.