Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-(Phenylcarbamoyl)benzoate 98.0+%, TCI America™
SDP

Catalog No. M26375G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M26375G Supplier TCI America Supplier No. M26375G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3814-10-6
Molecular Formula C15H13NO3
Molecular Weight (g/mol) 255.27
MDL Number MFCD00157832
InChI Key XTAGEOVGQDJRIC-UHFFFAOYSA-N
Synonym 4-(Phenylcarbamoyl)benzoic Acid Methyl Ester
PubChem CID 4682178
IUPAC Name methyl 4-(phenylcarbamoyl)benzoate
SMILES COC(=O)C1=CC=C(C=C1)C(=O)NC2=CC=CC=C2

Specifications

Melting Point 194°C
Color White
Quantity 5 g
Formula Weight 255.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-(Phenylcarbamoyl)benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.