Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-trifluoromethylbenzoylacetate, 95%, Thermo Scientific™

Catalog No. AC425440025
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC425440025 2.5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC425440025 Supplier Thermo Scientific Chemicals Supplier No. 425440025
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 212755-76-5
Molecular Formula C11H9F3O3
Molecular Weight (g/mol) 246.19
MDL Number MFCD00216521
InChI Key NMJOILIYLOHXGP-UHFFFAOYSA-N
Synonym methyl 4-trifluoromethyl benzoylacetate, methyl 4-trifluoromethylbenzoylacetate, methyl 3-oxo-3-4-trifluoromethyl phenyl propanoate, 3-oxo-3-4-trifluoromethylphenyl propionic acid methyl ester, methyl 3-4-trifluoromethyl phenyl-3-oxopropanoate, pubchem2696, buttpark 18\10-41, methyl 4-trifluoromethyl benzoyl acetate
PubChem CID 735882
IUPAC Name methyl 3-oxo-3-[4-(trifluoromethyl)phenyl]propanoate
SMILES COC(=O)CC(=O)C1=CC=C(C=C1)C(F)(F)F

Specifications

Melting Point 57°C to 62°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 2.5 g
Solubility Information Solubility in water: neglegible in water
Formula Weight 246.19
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-trifluoromethylbenzoylacetate, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.