Learn More
Medchemexpress LLC α-methyl-5-(2-thienylmethoxy)-1H-indole-3-ethanamine | 160521-72-2 | MFCD01321066 | 99.8% | 322.85 g/mol | C16H19ClN2OS | 5 MG
SDP
Catalog No. 5000213042
Encompass_Preferred
BW-723C86 is an orally active, selective 5-HT2B receptor agonist used in preclinical research to study serotonin-mediated effects. It has demonstrated anxiolytic-like activity and alters feeding, grooming, pulmonary, and gastric responses in animal models. The compound is supplied as a high-purity solid for laboratory use.
  • Selective 5-HT2B receptor agonist for receptor pharmacology studies.
  • Orally active, suitable for in vivo administration in animal models.
  • Reported anxiolytic-like and feeding-modulatory effects in preclinical assays.
  • High purity (~99.8%) appropriate for research applications.
  • Provided as a solid in small pack sizes for preclinical experiments.
  • Defined molecular formula and CAS number for unambiguous identification.
Catalog No. 50-002-13042 Supplier Medchemexpress LLC Supplier No. HY1013695MG
BW-723C86 is an orally active, selective 5-HT2B receptor agonist used in preclinical research to study serotonin-mediated effects. It has demonstrated anxiolytic-like activity and alters feeding, grooming, pulmonary, and gastric responses in animal models. The compound is supplied as a high-purity solid for laboratory use.
  • Selective 5-HT2B receptor agonist for receptor pharmacology studies.
  • Orally active, suitable for in vivo administration in animal models.
  • Reported anxiolytic-like and feeding-modulatory effects in preclinical assays.
  • High purity (~99.8%) appropriate for research applications.
  • Provided as a solid in small pack sizes for preclinical experiments.
  • Defined molecular formula and CAS number for unambiguous identification.