Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 5-bromo-2-furoate, 98%

Catalog No. AC355750010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC355750010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC355750010 Supplier Thermo Scientific Chemicals Supplier No. 355750010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2527-99-3 | C6H5BrO3 | 205.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2527-99-3
Molecular Formula C6H5BrO3
Molecular Weight (g/mol) 205.01
MDL Number MFCD00092313
InChI Key FBPIDMAELBIRLE-UHFFFAOYSA-N
Synonym methyl 5-bromo-2-furoate, methyl 5-bromo-2-furancarboxylate, 5-bromo-2-furoic acid methyl ester, 2-furancarboxylic acid, 5-bromo-, methyl ester, 5-bromo-2-furancarboxylic acid methyl ester, 5-bromofuran-2-carboxylic acid methyl ester, methyl 5-bromofuroate, 2-furoic acid, 5-bromo-, methyl ester, methyl 5-bromo-furanate, maybridge1_004996
PubChem CID 599633
IUPAC Name methyl 5-bromofuran-2-carboxylate
SMILES COC(=O)C1=CC=C(O1)Br

Specifications

Melting Point 65.0°C to 67.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 205.01
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Methyl 5-bromo-2-furoate, 98%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wash face,ha

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.