Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl 5-Doxyl-Stearate, Free Radical

Catalog No. 25361810MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-361-810MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 25-361-810MG Supplier Sigma Aldrich Supplier No. 25361810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Spin labelReactant free radical spin label involved in:Synthesis of alkyl phospholipid analogs of perifosine and miltefosine for cytotoxicity studiesStudies of micellular properties via electron spin (paramagnetic) resonanceSpin-labeled stearates partitioning into the lipid domain of stratum corneumLocation determinations in sodium dodecyl sulfate micellesStudies of local mobility and structure of micelles via its use as a probeESR (EPR) studies of metalloporphyrin intercalation into liposome membranes

Specifications

CAS 12075-68-2
Melting Point -50°C
Density 1.092 g/mL at 25°C
Molecular Formula C6H15Al2Cl3
Linear Formula (C2H5)3Al2Cl3
Quantity 10 mg
MDL Number MFCD00044852
Molecular Weight (g/mol) 247.51 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.