Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 5-Hydroxynicotinate 98.0+%, TCI America™
SDP

Catalog No. H08411G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08411G 1 g
H08415G 5 g
H084125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H08411G Supplier TCI America Supplier No. H08411G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 30766-22-4
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00191521
InChI Key KJJSHOHQQHACLE-UHFFFAOYSA-N
Synonym methyl 5-hydroxynicotinate, 5-hydroxynicotinic acid methyl ester, methyl5-hydroxynicotinate, methyl 5-hydroxy-3-pyridinecarboxylate, 5-hydroxy-nicotinic acid methyl ester, methyl-5-hydroxynicotinate, 5-hydroxy-3-pyridinecarboxylic acid methyl ester, 3-pyridinecarboxylic acid, 5-hydroxy-, methyl ester, 5-hydroxy nicotinic acid methyl ester, acmc-1crme
PubChem CID 354317
IUPAC Name methyl 5-hydroxypyridine-3-carboxylate
SMILES COC(=O)C1=CC(O)=CN=C1

Specifications

Melting Point 190°C
Color White
Quantity 1 g
Formula Weight 153.14
Percent Purity ≥98.0% (GC,T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material Methyl 5-Hydroxynicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.